Learn More
MYL3 Mouse anti-Human, CoraLite 594, Clone: 2E4B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66286-100UL
Description
The MYL3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MYL3. This antibody recognizes human, rat, mouse, pig antigen. The MYL3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| MYL3 | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 4634 | |
| IgG1 |
| Immunofluorescence | |
| 2E4B2 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |