missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYL12A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597190
This item is not returnable.
View return policy
Description
Immunogen sequence: KLNGTDPEDV IRNAFACFDE EATGTIQEDY LRELLTTMGD RFTDEEVDEL YREAPIDKKG NFNYIEFTRI LKHGAKDKDD; Positive Samples: LO2, A375, HeLa, K-562, Jurkat.
This gene encodes a nonsarcomeric myosin regulatory light chain. This protein is activated by phosphorylation and regulates smooth muscle and non-muscle cell contraction. This protein may also be involved in DNA damage repair by sequestering the transcriptional regulator apoptosis-antagonizing transcription factor (AATF)/Che-1 which functions as a repressor of p53-driven apoptosis. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 8.
Specifications
| MYL12A | |
| Polyclonal | |
| Unconjugated | |
| Myl12a | |
| 2900073G15Rik; AI838292; brain specific myosin regulatory light chain; C80200; Epididymis secretory protein Li 24; HEL-S-24; MLC-2; MLC20; MLC-2A; MLC-2B; MLCB; MRCL3; MRLC2; MRLC3; MYL12A; MYL12B; MYL2B; MYLC2B; myosin light chain 12A; myosin light chain, regulatory B-like; myosin regulatory light chain 12A; Myosin regulatory light chain 12B; Myosin regulatory light chain 2, nonsarcomeric; Myosin regulatory light chain 20 kDa; myosin regulatory light chain 2-A, smooth muscle isoform; Myosin regulatory light chain 2-B, smooth muscle isoform; myosin regulatory light chain 3; Myosin regulatory light chain MRLC2; Myosin regulatory light chain MRLC3; myosin regulatory light chain RLC-A; Myosin RLC; myosin RLC-A; myosin, light chain 12A, regulatory, non-sarcomeric; myosin, light polypeptide, regulatory, non-sarcomeric (20kD); NMDA receptor-interacting protein; RGD1309537; RLC; Rlc-a; SHUJUN-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10627 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.87 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P19105 | |
| Myl12a | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 92-171 of human MYL12A (NP_0064621). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction