missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15814-1-AP-150UL
This item is not returnable.
View return policy
Description
Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in fast skeletal muscle. Two transcript variants have been identified for this gene.Specifications
| MYL1 | |
| Polyclonal | |
| Unconjugated | |
| MYL1 | |
| MLC1/MLC3, MLC1F, MLC1F/MLC3F, MLC3F, MYL1, Myosin light chain A1/A2, Myosin light chain alkali 1/2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17901, 4632, 56781 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02600, P05976, P05977 | |
| Myl1 | |
| MYL1 Fusion Protein Ag8594 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction