Learn More
Invitrogen™ Myosin Skeletal Muscle Monoclonal Antibody (MY32)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA190701
Description
This antibody recognizes heavy chain human skeletal myosin (fast-twitch type II isomyosin). It does not stain cardiac or smooth muscle myosin. For antigen retrieval, it is recommended to use a citrate buffer, pH 6.0, at room temperature for 30 min. Store as a concentrated solution. Centrifuge briefly prior to opening vial. Store at 4°C.
Myosin is a major contractile protein which converts chemical energy into mechanical energy through the hydrolysis of ATP. Myosin is a hexameric protein composed of a pair of myosin heavy chains (MYH) and two pairs of nonidentical light chains. Myosin heavy chains are encoded by a multigene family. In mammals at least 10 different myosin heavy chain (MYH) isoforms have been described from striated, smooth, and nonmuscle cells. These isoforms show expression that is spatially and temporally regulated during development.
Specifications
| Myosin Skeletal Muscle | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.09% sodium azide | |
| P12882, Q28641, Q29RW1, Q9UKX2, Q9Y623 | |
| Myh1, MYH2, MYH4 | |
| Rabbit muscle myosin. | |
| 6 mL | |
| Primary | |
| Human, Rat, Rabbit | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| MY32 | |
| Unconjugated | |
| Myh1 | |
| 2A myosin heavy chain; 2B myosin heavy chain; A530084A17Rik; AI506973; epididymis luminal protein 71; fast 2a myosin heavy chain; fast myosin heavy chain; fast myosin heavy chain 2X; HEL71; IBM3; IId; IId/x; inclusion body myopathy 3, autosomal dominant; MdMs; MGC133384; MHC; MHC2A; MHC2B; MHC2X/D; MHC-2X/D; Minimsc; Minmus; MM; MYH1; MYH2; MYH2A; MYH2B; MYH-2B; Myh4; MYHa; MYHas8; MYHC; MyHC-2A; MyHC-2b; MYHC-2d; MyHC-2x; MyHC-2X/D; MyHC-IIA; MyHC-IIb; MyHC-IId/x; MYHC-IIX; MyHC-IIx/d; MYHSA1; MYHSA2; Myhsf; Myhs-f; Myhsf1; Myhs-f1; Myhsf2; Myhs-f2; myosin heavy chain; myosin heavy chain 1; Myosin heavy chain 2; myosin heavy chain 2a; Myosin heavy chain 2b; myosin heavy chain 2d; myosin heavy chain 2X; Myosin heavy chain 4; myosin heavy chain IIa; Myosin heavy chain IIb; myosin heavy chain IIx/d; myosin heavy chain isoform 2b; myosin heavy chain isoform fast type; myosin heavy chain type IIb; myosin heavy chain type IIx; myosin heavy chain, skeletal muscle; myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain, skeletal muscle, adult 2; myosin heavy chain, skeletal muscle, fetal; Myosin heavy chain, skeletal muscle, juvenile; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 2, skeletal muscle, adult; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 2, skeletal muscle, adult; myosin, heavy polypeptide 4, skeletal muscle; myosin-1; Myosin-2; myosin-4; MYPOP; skeletal muscle myosin heavy chain II-a; type 2X myosin heavy chain; type IIA myosin heavy chain; unnamed protein product | |
| Mouse | |
| IgG fraction | |
| RUO | |
| 100009054, 100009548, 100125991, 287408, 360543, 4619, 4620, 4622, 691644 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.