missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH8 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41309AF750
This item is not returnable.
View return policy
Description
MYH8 Polyclonal antibody specifically detects MYH8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| MYH8 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 4626 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| Fetal-Myosin Heavy Chain, gtMHC-F, MyHC-peri, myHC-perinatal, MyHC-pn, Myosin Heavy Chain 8, Myosin Heavy Chain, Skeletal Muscle, Perinatal, Myosin, Heavy Chain 8, Skeletal Muscle, Perinatal, Myosin, Heavy Polypeptide 8, Skeletal Muscle, Perinatal, myosin-8 | |
| Antibody was raised against a 19 amino acid peptide near the amino terminus of human MYH8. The immunogen is located within amino acids 60 - 110 of MYH8. Amino Acid Squence: VREDQVFPMNPPKYDKIED | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction