missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYH7B Recombinant Rabbit Monoclonal Antibody (JE54-44)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA536186
This item is not returnable.
View return policy
Description
MYH7B Recombinant Monoclonal Antibody for Western Blot
Myosin cardiac muscle beta chain or MYH7B is believed to be involved in cardiac slow-twitch myosin contraction.
Specifications
| MYH7B | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| A2AQP0, A7E2Y1 | |
| MYH7B | |
| Recombinant protein within Human MYH7B aa 1,710-1,983. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| JE54-44 | |
| Unconjugated | |
| MYH7B | |
| A830009F23Rik; AA517445; alpha cardiac MHC; alpha myosin; alpha-cardiac myosin heavy chain; alpha-MHC; Antigen MLAA-21; ASD3; beta cardiac myosin heavy chain; beta myosin heavy chain; beta-MHC; beta-myosin heavy chain; B-MHC; cardiac muscle myosin heavy chain 7 beta; cardiac myosin heavy chain alpha isoform; cardiac myosin heavy chain beta isoform; cardiomyopathy, hypertrophic 1; CMD1EE; CMD1S; CMH1; CMH14; KIAA1512; LOC100736765; MHC14; MPD1; Myh14; MYH6; MYH7; Myh7b; MYH-beta/slow; MYHC; Myhca; Myhc-a; MyHC-alpha; Myhcb; Myhc-b; MyHC-beta; MyHC-I; myhc-slow; myopathy, distal 1; myosin 7; Myosin cardiac muscle beta chain; Myosin heavy chain 6; myosin heavy chain 7; myosin heavy chain 7B; myosin heavy chain 7B, cardiac muscle beta isoform; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain polypeptide 7 cardiac muscle fetal; myosin heavy chain slow; myosin heavy chain slow isoform; myosin heavy chain slow type 1 (beta cardiac); myosin heavy chain, cardiac; Myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle beta isoform; myosin heavy chain, cardiac muscle, adult; myosin heavy chain, cardiac muscle, fetal; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin heavy chain, polypeptide 7; myosin, heavy chain 6, cardiac muscle, alpha; myosin, heavy chain 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin, heavy chain 7, cardiac muscle, beta; myosin, heavy chain 7B, cardiac muscle, beta; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin, heavy polypeptide 7, cardiac muscle, beta; myosin, heavy polypeptide 7B, cardiac muscle, beta; myosin-6; Myosin-7; myosin-7B; rhabdomyosarcoma antigen MU-RMS-40.7A; Slow A MYH14; slow myosin heavy chain-beta; SPMD; SPMM; SSS3; type slow/beta myosin heavy chain; U937-associated antigen; Unknown (protein for IMAGE:7985957) | |
| Rabbit | |
| Protein A | |
| RUO | |
| 311570, 57644, 668940 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction