missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH6 Antibody (CL2148) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44471
This item is not returnable.
View return policy
Description
MYH6 Monoclonal antibody specifically detects MYH6 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
Specifications
| MYH6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
| alpha-MHC, ASD3, cardiomyopathy, CMD1EE, CMH14, Hypertrophic 1, MYHC, MYHCA, myHC-alpha, Myosin Heavy Chain 6, Myosin Heavy Chain, Cardiac Muscle Alpha Isoform, Myosin, Heavy Chain 6, Cardiac Muscle, Alpha, Myosin, Heavy Polypeptide 6, Cardiac Muscle, Alpha (Cardiomyopathy, Hypertrophic 1), myosin-6, SSS3 | |
| This antibody was generated using a recombinant protein sequence of P13533, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Cell Biology, Cellular Markers, Hypoxia, Stem Cell Markers | |
| 4624 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| CL2148 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction