missing translation for 'onlineSavingsMsg'
Learn More

MYH6/7 Rabbit anti-Mouse, Rat, Clone: 6V0G1, Novus Biologicals™

Product Code. 18364792 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364792 100 μg 100µL
18385904 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18364792 Supplier Novus Biologicals Supplier No. NBP316520100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MYH6/7 Monoclonal antibody specifically detects MYH6/7 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH6/7
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6V0G1
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ASD3, CMD1EE, CMD1S, CMH1, CMH14, MPD1, MYHC, myHC-alpha, MyHC-beta, myhc-slow, Myosin Heavy Chain 6, Myosin heavy chain 7, Myosin heavy chain slow isoform, Myosin Heavy Chain, Cardiac Muscle Alpha Isoform, Myosin heavy chain, cardiac muscle beta isoform, myosin-6, myosin-7, SPMD, SSS3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1200-1400 of human MYH6/7 (P12883).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4624
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.