missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23357
This item is not returnable.
View return policy
Description
MYH4 Polyclonal antibody specifically detects MYH4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MYH4 | |
| Polyclonal | |
| Western Blot 1:2000 | |
| MyHC-2B, MyHC-IIb, myosin heavy chain 4, myosin heavy chain IIb, myosin heavy chain, skeletal muscle, fetal, myosin, heavy chain 4, skeletal muscle, myosin, heavy polypeptide 4, skeletal muscle, myosin-4 | |
| A synthetic peptide (coupled to KLH) corresponding to amino acid residues at the C-terminus from mouse MYH4. This sequence has 100% identity with rat and human MYH4, and has significant homology to MYH1, MYH2, and MYH8. [UniProt# Q5SX39] | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 4622 | |
| Store at -20°C. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction