missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55069-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to MYH2, will not bind other myosins.
Muscle contraction. Required for cytoskeleton organization.Specifications
| MYH2 | |
| Polyclonal | |
| Unconjugated | |
| MYH2 | |
| MYH2, MYH2A, MYHas8, MyHC 2A, MyHC IIa, MYHSA2, Myosin 2, Myosin 2a, Myosin heavy chain 2, Myosin heavy chain 2a, Myosin heavy chain IIa | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4620 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.26 mg/mL | |
| 0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UKX2 | |
| MYH2 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction