Learn More
MYH15 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22290-1-AP-20UL
This item is not returnable.
View return policy
Description
The MYH15 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human MYH15. This antibody recognizes human, mouse antigen. The MYH15 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| MYH15 | |
| 650 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| KIAA1000, MYH15, Myosin 15, Myosin heavy chain 15, myosin, heavy chain 15 | |
| 20 μL | |
| 22989 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |