missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYH14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564436
This item is not returnable.
View return policy
Description
Immunogen sequence: GAGEQLKADL LLEPCSHYRF LTNGPSSSPG QERELFQETL ESLRVLGFSH EE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.
Cellular myosin that appears to play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping.
Specifications
| MYH14 | |
| Polyclonal | |
| Unconjugated | |
| MYH14 | |
| 2400004E04Rik; DFNA4; DFNA4A; FP17425; II-C; KIAA2034; MHC16; Myh14; MYH14 variant protein; MYH17; myosin; myosin heavy chain 14; Myosin heavy chain, non-muscle IIc; myosin, heavy chain 14; myosin, heavy chain 14, non-muscle; myosin, heavy polypeptide 14; myosin-14; NHMCII; NMHC II-C; NMHC-II-C; nonmuscle myosin heavy chain; Non-muscle myosin heavy chain IIc; nonmuscle myosin heavy chain II-C; PNMHH | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79784 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z406 | |
| MYH14 | |
| Recombinant protein fragment of Human MYH14 (Product #RP-104174) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction