Learn More
MYH10-Specific Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-19673-20UL
Description
The MYH10-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human MYH10-Specific. This antibody recognizes human, mouse, rat antigen. The MYH10-Specific antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| MYH10-Specific | |
| 1000 μg/mL | |
| MYH10, Myosin 10, Myosin heavy chain 10 | |
| 20 μL | |
| 4628 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.