missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21403-1-AP-150UL
This item is not returnable.
View return policy
Description
MYH10 encodes a member of the myosin superfamily. The protein represents a conventional non-muscle myosin; it should not be confused with the unconventional myosin-10 (MYO10). Myosins are actin-dependent motor proteins with diverse functions including regulation of cytokinesis, cell motility, and cell polarity. Mutations in this gene have been associated with May-Hegglin anomaly and developmental defects in brain and heart. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| MYH10 | |
| Polyclonal | |
| Unconjugated | |
| MYH10 | |
| MYH10, Myosin 10, Myosin heavy chain 10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4628, 77579, 79433 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P35580, Q61879, Q9JLT0 | |
| MYH10 | |
| MYH10 Fusion Protein Ag16113 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction