missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYH10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587092
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
MYH10 encodes a member of the myosin superfamily. The protein represents a conventional non-muscle myosin; it should not be confused with the unconventional myosin-10 (MYO10). Myosins are actin-dependent motor proteins with diverse functions including regulation of cytokinesis, cell motility, and cell polarity. Mutations in this gene have been associated with May-Hegglin anomaly and developmental defects in brain and heart. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| MYH10 | |
| Polyclonal | |
| Unconjugated | |
| MYH10 | |
| 5730504C04Rik; 9330167F11Rik; cellular myosin heavy chain, type B; flectin; Fltn; MCH-B; MCH-B(B2); mKIAA3005; MYH10; Myhn2; Myhn-2; Myosin heavy chain 10; myosin heavy chain 10, non-muscle; myosin heavy chain, non-muscle IIb; myosin heavy chain, nonmuscle type B; myosin heavy chain, nonmuscular, type B; myosin IIB; myosin, heavy chain 10, non-muscle; myosin, heavy polypeptide 10, non-muscle; myosin-10; NMHC II-B; NMHC-B; NMHCII-B; NMMHC IIB; NMMHC II-b; NMMHCB; NMMHC-B; NMMHC-IIB; nonmuscle myosin; Non-muscle myosin heavy chain B; nonmuscle myosin heavy chain IIB; non-muscle myosin heavy chain IIb; nonmuscle myosin heavy chain II-B; nonmuscle myosin heavy chain-B; nonmuscle myosin II heavy chain-B; OTTHUMP00000165799; SMemb | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4628, 77579, 79433 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P35580, Q61879, Q9JLT0 | |
| MYH10 | |
| Recombinant fusion protein of human MYH10. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction