missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MYH Polyclonal antibody specifically detects MYH in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
Specifications
| Antigen | MYH |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.2.2, EC 3.2.2.-, EC 3.2.2.31, hMYH, MGC4416, MutY homolog, MUTYH, Muyth, MYH, MYHA/G-specific adenine DNA glycosylase |
| Host Species | Rabbit |
| Immunogen | mutY homolog(E. coli). (Uniprot# Q9UIF7) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?