Learn More
Invitrogen™ MYF6 Monoclonal Antibody (5D7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517131
Description
MA5-17131 targets MYF6 in indirect ELISA and WB applications and shows reactivity with Human samples. The MA5-17131 immunogen is purified recombinant fragment of human MYF6 (amino acids: 85-150) expressed in E. Coli. MA5-17131 detects MYF6 which has a predicted molecular weight of approximately 27kDa.
Involved in muscle differentiation (myogenic factor). Induces fibroblasts to differentiate into myoblasts. Probable sequence specific DNA-binding protein.
Specifications
| MYF6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P23409 | |
| Myf6 | |
| Purified recombinant fragment of human MYF6 (amino acids: 85-150) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 5D7 | |
| Unconjugated | |
| Myf6 | |
| bHLHc4; Class C basic helix-loop-helix protein 4; CNM3; Herculin; MRF4; MRF4AA; Muscle-specific regulatory factor 4; Myf6; myf-6; Myogenic factor 6; Myogenic factor 6 (herculin); myogenic factor 6 (herculin); myogenic factor-6 | |
| Mouse | |
| Protein G | |
| RUO | |
| 4618 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.