missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYF-5 Antibody (OTI2G5), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72855PE
This item is not returnable.
View return policy
Description
MYF-5 Monoclonal antibody specifically detects MYF-5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MYF-5 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 4617 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2G5 | |
| Western Blot, Immunohistochemistry | |
| BHLHC2, bHLHc2Class C basic helix-loop-helix protein 2, myf-5, myogenic factor 5 | |
| Full length human recombinant protein of human MYF5 (NP_005584) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction