missing translation for 'onlineSavingsMsg'
Learn More

MYEOV Antibody [DyLight 405], Novus Biologicals™

Product Code. 30764057 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30764057 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30764057 Supplier Novus Biologicals Supplier No. NBP346986V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MYEOV Polyclonal antibody specifically detects MYEOV in Human,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYEOV
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias myeloma overexpressed (in a subset of t(11;14) positive multiple myelomas), myeloma overexpressed gene (in a subset of t(11;14) positive multiple myelomas), OCIMhypothetical protein LOC26579, Oncogene in multiple myeloma
Host Species Rabbit
Immunogen myeloma overexpressed(in a subset of t(11;14) positive multiple myelomas). (Uniprot# Q96EZ4)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26579
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.