missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloperoxidase/MPO Antibody (MPO/8997R), Novus Biologicals™
Rabbit Monoclonal Antibody
£214.00 - £377.00
Specifications
| Antigen | Myeloperoxidase/MPO |
|---|---|
| Clone | MPO/8997R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18664154
|
Novus Biologicals
NBP3-23860-100ug |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18629663
|
Novus Biologicals
NBP3-23860-20ug |
20 μg |
£214.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifica
| Myeloperoxidase/MPO | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism | |
| 10 mM PBS with 0.05% BSA | |
| 4353 | |
| IgG κ | |
| Protein A or G purified |
| MPO/8997R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.11.1, EC 1.11.1.7, myeloperoxidase | |
| Recombinant fragment of human Myeloperoxidase/MPO protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto