Learn More
Myeloperoxidase/MPO Antibody (CL15002) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26847AF647
Product Code. 30301051
  • £558.00 / 0.10mL
Description
Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Myeloperoxidase/MPO
Monoclonal
Alexa Fluor 647
EC 1.11.1, EC 1.11.1.7, myeloperoxidase
This antibody was developed using a synthetic peptide derived from P05164, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
4353
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot
CL15002
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified