missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloperoxidase/MPO Antibody (CL14976) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26845F
This item is not returnable.
View return policy
Description
Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Myeloperoxidase/MPO | |
| Monoclonal | |
| FITC | |
| EC 1.11.1, EC 1.11.1.7, myeloperoxidase | |
| This antibody was developed using a synthetic peptide derived from P05164, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism | |
| 4353 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot | |
| CL14976 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction