missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Myeloperoxidase Monoclonal Antibody (8E6), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ GM4192
This item is not returnable.
View return policy
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
Myeloperoxidase (MPO) is a heme protein that is synthesized during myeloid differentiation and is a major component of neutrophil azurophilic granules. It is produced as a single chain precursor and subsequently cleaved into a light and heavy chain. The mature MPO is a tetramer composed of 2 light chains and 2 heavy chains. This enzyme is responsible for producing hypohalous acids that are central to the microbicidal activity of neutrophils. Diseases that are associated with MPO include Myeloperoxidase Deficiency and Alzheimer's Disease, Familial, 1.
Specifications
| Myeloperoxidase | |
| Monoclonal | |
| FITC | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.2 | |
| P05164 | |
| MPO | |
| Human Myeloperoxidase. | |
| 1 mL | |
| Primary | |
| 1 Each | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 8E6 | |
| Assay-dependent | |
| MPO | |
| 84 kDa myeloperoxidase; 89 kDa myeloperoxidase; mKIAA4033; MPO; Myeloperoxidase; Myeloperoxidase heavy chain; Myeloperoxidase light chain | |
| Mouse | |
| purified | |
| ASR | |
| 4353 | |
| 0.05% Sodium azide | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction