missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Myeloid Specific Antigen Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101681
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MNDA.
This product provides a new panel of monoclonal antibodies which stain early precursor and mature forms of human myeloid cells. This panel of monoclonal antibodies reacts with antigenic determinants present in normal myeloid cells and leukemias of similar derivation.
Specifications
| Myeloid Specific Antigen | |
| Polyclonal | |
| Unconjugated | |
| MNDA | |
| Ifi204; interferon activated gene 204; MNDA; Myeloid cell nuclear differentiation antigen; PYHIN3; rHin-3; SB135 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4332 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P41218 | |
| MNDA | |
| A synthesized peptide derived from human MNDA(Accession P41218), corresponding to amino acid residues L122-H172. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction