missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Myeloid Specific Antigen Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575418
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This product provides a new panel of monoclonal antibodies which stain early precursor and mature forms of human myeloid cells. This panel of monoclonal antibodies reacts with antigenic determinants present in normal myeloid cells and leukemias of similar derivation.
Specifications
| Myeloid Specific Antigen | |
| Polyclonal | |
| Unconjugated | |
| MNDA | |
| Ifi204; interferon activated gene 204; MNDA; Myeloid cell nuclear differentiation antigen; PYHIN3; rHin-3; SB135 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 304988, 4332 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P41218 | |
| MNDA | |
| Synthetic peptide corresponding to amino acids 361-407 of Human MNDA. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction