missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloid leukemia factor 1 Antibody (OTI3F11), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72849AF405
This item is not returnable.
View return policy
Description
Myeloid leukemia factor 1 Monoclonal specifically detects Myeloid leukemia factor 1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Myeloid leukemia factor 1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MLF1 | |
| Full length recombinant protein of human Myeloid leukemia factor 1 (NP_071888) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI3F11 | |
| Western Blot, Immunohistochemistry | |
| Myelodysplasia-myeloid leukemia factor 1, myeloid leukemia factor 1 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Stem Cell Markers | |
| 4291.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction