missing translation for 'onlineSavingsMsg'
Learn More

Myeloid Cell Marker Antibody (BM-2), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18380256 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380256 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380256 Supplier Novus Biologicals Supplier No. NBP314086AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myeloid Cell Marker Monoclonal antibody specifically detects Myeloid Cell Marker in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myeloid Cell Marker
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BM-2
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias myeloid specific antigen, myeloid specific protein, p183 DNA-binding cytoplasmic ptotein
Host Species Mouse
Immunogen Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.