missing translation for 'onlineSavingsMsg'
Learn More

Myeloid-Associated Differentiation Marker Antibody (MYADM/972), PE, Novus Biologicals™

Product Code. 18628066 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18628066 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18628066 Supplier Novus Biologicals Supplier No. NBP311468PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myeloid-Associated Differentiation Marker Monoclonal antibody specifically detects Myeloid-Associated Differentiation Marker in Human, Macaque samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myeloid-Associated Differentiation Marker
Applications Flow Cytometry
Classification Monoclonal
Clone MYADM/972
Conjugate PE
Dilution Flow Cytometry
Formulation PBS
Gene Alias MYADM, Myeloid Upregulated Protein, Protein SB135, SB135
Host Species Mouse
Immunogen Recombinant human Myeloid-Associated Differentiation Marker protein (Uniprot: Q96S97)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91663
Target Species Human, Macaque
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.