missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloid-Associated Differentiation Marker Antibody (MYADM/971), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11489PCP
This item is not returnable.
View return policy
Description
Myeloid-Associated Differentiation Marker Monoclonal antibody specifically detects Myeloid-Associated Differentiation Marker in Human, Macaque samples. It is validated for Flow Cytometry
Specifications
| Myeloid-Associated Differentiation Marker | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 91663 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| MYADM/971 | |
| Flow Cytometry | |
| MYADM, Myeloid Upregulated Protein, Protein SB135, SB135 | |
| Recombinant full-length human Myeloid-Associated Differentiation Marker protein. (Uniprot: Q96S97) | |
| 0.1 mL | |
| Primary | |
| Human, Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction