missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloid-Associated Differentiation Marker Antibody (MYADM/971) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Myeloid-Associated Differentiation Marker Monoclonal antibody specifically detects Myeloid-Associated Differentiation Marker in Human, Monkey, Macaque samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Specifications
Specifications
| Antigen | Myeloid-Associated Differentiation Marker |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Clone | MYADM/971 |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml |
| Formulation | 10 mM PBS |
| Gene Alias | MYADM, Myeloid Upregulated Protein, Protein SB135, SB135 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?