missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies MyeloCult™ M5300

Product Code. 17168261
Encompass
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17168261 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17168261 Supplier STEMCELL Technologies Supplier No. 05350

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Medium for long-term culture of mouse cells

  • MyeloCult™ M5300 is a culture medium for the initiation and maintenance of myeloid long-term cultures of mouse hematopoietic cells and stromal cell feeder layers
  • The sera used in this formulation have been pre-tested and selected for their ability to support long-term myelopoiesis by primitive mouse hematopoietic cells (e.g. in long-term culture-initiating cell [LTC-IC] assays)
  • MyeloCult™ M5300 requires the addition of freshly prepared Hydrocortisone (Catalog #07904)

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Culture, Functional Assay
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.