missing translation for 'onlineSavingsMsg'
Learn More

Myelin PLP Mouse anti-Human, Mouse, Rat, Bovine, Reptile, Clone: PLP1/4259, Novus Biologicals™

Product Code. 18393743 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393743 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393743 Supplier Novus Biologicals Supplier No. NBP30760320UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myelin PLP Monoclonal specifically detects Myelin PLP in Human, Mouse, Rat, Bovine, Reptile samples. It is validated for Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myelin PLP
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PLP1/4259
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias HLD1, lipophilin, major myelin proteolipid protein, MMPL, myelin proteolipid protein, PLP, PLP/DM20, PMD, proteolipid protein 1, spastic paraplegia 2, uncomplicated, SPG2
Host Species Mouse
Immunogen Synthetic peptide GRGTKF that recognizes the C terminal region (aa272-277) of Myelin PLP.
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5354
Target Species Human, Mouse, Rat, Bovine, Reptile
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.