Learn More
Myelin PLP Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB100-1608
This item is not returnable.
View return policy
Description
Myelin PLP Polyclonal specifically detects Myelin PLP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Western Blot, Immunocytochemistry, Immunohistochemistry (Frozen) | |
| PLP1 | |
| Affinity Purified | |
| RUO |
| Polyclonal | |
| Chickens were immunized with a synthetic peptide/keyhole limpet hemocyanin (KLH) conjugate. The peptide corresponded to a region of the PLP gene product shared between the human (NP_000524, PLP1 isoform 1 and PLP1 isoform 2 (NP_955772)] and mouse (P60202). After | |
| 0.25 mL | |
| Store at 4C in the dark. |