missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myelin expression factor 2 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-12765PCP
This item is not returnable.
View return policy
Description
Myelin expression factor 2 Polyclonal antibody specifically detects Myelin expression factor 2 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Myelin expression factor 2 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| FLJ11213, HsT18564, KIAA1341MSTP156, MEF-2myEF-2, MGC87325, MST156, MyEF-2, myelin expression factor 2, myelin gene expression factor 2 | |
| The immunogen this antibody was made to, maps to a region between residue 25 and 50 of human Myelin Expression Factor 2 using the numbering given in entry NP_057216.2 (GeneID 50804). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 50804 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction