missing translation for 'onlineSavingsMsg'
Learn More

Myelin expression factor 2 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18426385 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426385 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426385 Supplier Novus Biologicals Supplier No. NBP212765C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Myelin expression factor 2 Polyclonal antibody specifically detects Myelin expression factor 2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myelin expression factor 2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ11213, HsT18564, KIAA1341MSTP156, MEF-2myEF-2, MGC87325, MST156, MyEF-2, myelin expression factor 2, myelin gene expression factor 2
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 25 and 50 of human Myelin Expression Factor 2 using the numbering given in entry NP_057216.2 (GeneID 50804).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50804
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.