missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MyD88 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529611
This item is not returnable.
View return policy
Description
Recommended positive controls: MyD88-transfected 293T. Predicted reactivity: Mouse (82%), Rat (81%), Pig (86%), Sheep (83%), Rhesus Monkey (99%), Chimpanzee (99%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Myeloid differentiation Marker 88 (MyD88) is an adaptor molecule for the Toll/IL-1R and is involved in the inflammatory response induced by IL-1, IL-18 and LPS. MyD88 recruits IRAK to IL-1 receptor complex in response to IL-1. This pathway further leads to activation of NF-kB. MyD88 has a wide tissue distribution.
Specifications
| MyD88 | |
| Polyclonal | |
| Unconjugated | |
| MYD88 | |
| mutant myeloid differentiation primary response 88; Myd88; MYD88D; myeloid differentiation primary response 88; myeloid differentiation primary response gene (88); myeloid differentiation primary response gene 88; myeloid differentiation primary response protein MyD88 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17874, 4615 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| 1.18 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P22366, Q99836 | |
| MYD88 | |
| Recombinant protein encompassing a sequence within the center region of human MYD88. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction