Learn More
Invitrogen™ MyD88 Polyclonal Antibody

Description
Suggested positive controls are A431, A549, K562, HepG2, or NIH/3T3 Cell Lysates. PA5-19919 can be used with blocking peptide PEP-0045. Predicted molecular ~ 35kD. In Western blot applications, this antibody has been observed to detect a band at: 35kD Predicted species reactivity based on immunogen sequence: Pig: (94%), Sheep: (82%), Bovine: (82%), Chicken: (82%) Antibodies should not be exposed to prolonged high temperatures.
Specifications
Specifications
| Antigen | MyD88 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | MYD88 |
| Gene Accession No. | P22366, Q6Y1S1, Q99836 |
| Gene Alias | mutant myeloid differentiation primary response 88; Myd88; MYD88D; myeloid differentiation primary response 88; myeloid differentiation primary response gene (88); myeloid differentiation primary response gene 88; myeloid differentiation primary response protein MyD88 |
| Gene Symbols | MYD88 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.