missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyD88 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-76465CL1
This item is not returnable.
View return policy
Description
MyD88 Polyclonal antibody specifically detects MyD88 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot,KnockDown,Immunocytochemistry/Immunofluorescence
Specifications
| MyD88 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot, KnockDown, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 | |
| Antibody (2127) was raised against a peptide corresponding to 17 amino acids near carboxy terminus of human MYD88 isoform 1. The immunogen is located within the last 50 amino acids of MYD88. Amino Acid Squence: CTKSWFWTRLAKALSLP | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
| 4615 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction