missing translation for 'onlineSavingsMsg'
Learn More

MyD88 Antibody (4D6), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18400337 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400337 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400337 Supplier Novus Biologicals Supplier No. NBP227369JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MyD88 Monoclonal antibody specifically detects MyD88 in Human, Mouse, Primate, Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Flow (Intracellular), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen MyD88
Applications Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 4D6
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88
Host Species Mouse
Immunogen A portion of amino acids 50-100 of human MyD88 was used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4615
Target Species Human, Mouse, Primate, Macaque
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.