missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyD88 Antibody (4D6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80862
This item is not returnable.
View return policy
Description
MyD88 Monoclonal specifically detects MyD88 in Human, Mouse, Primate, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular), Immunocytochemistry, CyTOF-ready.
Specifications
| MyD88 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 5-7 μg/mL, Flow Cytometry : 0.5ug/10^6cells, Immunocytochemistry/Immunofluorescence : 5 μg/mL, Flow (Intracellular), Immunocytochemistry, CyTOF-ready | |
| MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Gorilla, Chimp, Orangutan (100%), Rat (63%), Equine (82%), Canine (73%), Cat, Mouse (68%). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF | |
| 4D6 | |
| Unconjugated | |
| PBS | |
| MYD88 | |
| A portion of amino acids 50-100 of human MyD88 was used as the immunogen for this antibody. | |
| 0.1 mg | |
| Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
| 4615 | |
| Human, Mouse, Primate, Rhesus Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction