missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyD88 Antibody (316628) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB29281JF635
This item is not returnable.
View return policy
Description
MyD88 Monoclonal antibody specifically detects MyD88 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| MyD88 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| MYD88D, myeloid differentiation primary response gene (88), myeloid differentiation primary response protein MyD88 | |
| E. coli-derived recombinant human MyD88, Met1-Pro296, Accession # Q99836 | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
| 4615 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 316628 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction