missing translation for 'onlineSavingsMsg'
Learn More

MYCT1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18406956 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406956 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406956 Supplier Novus Biologicals Supplier No. NBP281921AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MYCT1 Polyclonal antibody specifically detects MYCT1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYCT1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias FLJ21269, MGC156309, MGC156310, MTLC, MTLCmyc target in myeloid cells 1, MTMC1, myc target 1, Myc target in myeloid cells protein 1, myc target protein 1
Host Species Rabbit
Immunogen MYCT1 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human MYCT1. The immunogen is located within the last 50 amino acids of MYCT1. Amino Acid Squence: TSHSLSRPDYWSSNSLR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80177
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.