missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mycoplasma pneumoniae Monoclonal Antibody (Q41F)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518235
This item is not returnable.
View return policy
Description
This antibody is reactive with 70 and 80 kDa WB bands of M. pneumoniae and does not react with uninfected cells. Purity is >90%.
Although they are the simplest and smallest self-replicating bacteria, Mycoplasma infections tend to be persistent, difficult to detect, diagnose, and cure. Mycoplasmas are surface parasites of human respiratory and urogenital tracts. Mycoplasma pneumoniae is often the causative agent of community-acquired pneumonia. The organism's affinity for respiratory epithelial cells and its ability to secrete hydrogen peroxide are a potent combination, leading to oxidative tissue damage.
Specifications
| Mycoplasma pneumoniae | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot, Immunocytochemistry | |
| Q41F | |
| Unconjugated | |
| Bacterial pneumonia; M. pneumoniae; mycoplasmen | |
| Mycoplasma pneumoniae native protein. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction