Learn More
Invitrogen™ Mycoplasma bovis Monoclonal Antibody (MY10-151.1), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA5-46155
 View more versions of this product
Product Code. 17800346
  • £916.00 / 1mg
Description
MA1-10792 detects Mycoplasma bovis. MA1-10792 has been successfully used in ELISA applications. The MA1-10792 immunogen is Mycoplasma bovis cells.
This antibody specifically recognize Mycoplasma bovis. It does not cross-react with Mycoplasma genitalium, Mycoplasma californicum, Strep. agalactiae, Staph. aureus or E. coli.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mycoplasma bovis
Monoclonal
1.5 mg/mL
PBS with no preservative; pH 7.4
Mouse
Protein A
RUO
Bacteria
Antibody
IgG2a
ELISA
MY10-151.1
Unconjugated
m bovis; M. bovis; mycoplasmen
Mycoplasma bovis cells.
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles