missing translation for 'onlineSavingsMsg'
Learn More

Mycophenolic acid-d3, MedChemExpress

Product Code. 16432169
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16432169 1 mg 1mg
16400928 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16432169 Supplier Medchem Express Supplier No. HYB0421S
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Mycophenolic acid D3 (Mycophenolate D3) is deuterium labeled Mycophenolic acid, which is an an immunosuppresant drug and has potent anti-proliferative activity.

Mycophenolic acid D3 (Mycophenolate D3) is deuterium labeled Mycophenolic acid, which is an an immunosuppresant drug and has potent anti-proliferative activity.

Specifications

Chemical Name or Material Mycophenolic acid-d3
CAS 1185242-90-3
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C17H17D3O6
Color White
Synonym Mycophenolate-d3
SMILES O=C(OC1)C2=C1C(C)=C(OC([2H])([2H])[2H])C(C/C=C(C)/CCC(O)=O)=C2O
Molecular Weight (g/mol) 323.36
Quantity 1 mg
Formula Weight 323.36
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.