missing translation for 'onlineSavingsMsg'
Learn More

Mycophenolic Acid Acyl-Beta-D-glucuronide, TRC

Product Code. 30137640
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137640 10 mg 10mg
30137623 5 mg 5mg
18003220 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137640 Supplier Toronto Research Chemicals Supplier No. TRCM83152510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99043-04-6
Molecular Formula C23 H28 O12
Molecular Weight (g/mol) 496.46
Synonym Mycophenolic Acid Acyl-B-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[(E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoyl]oxyoxane-2-carboxylic acid
SMILES COc1c(C)c2COC(=O)c2c(O)c1C\C=C(/C)\CCC(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H28O12/c1-9(4-6-11-15(25)14-12(8-33-22(14)31)10(2)19(11)32-3)5-7-13(24)34-23-18(28)16(26)17(27)20(35-23)21(29)30/h4,16-18,20,23,25-28H,5-8H2,1-3H3,(H,29,30)/b9-4+/t16-,17-,18+,20-,23+/m0/s1
Formula Weight 496.1581
Percent Purity >95%
Chemical Name or Material Mycophenolic acid acyl-beta-D-glucuronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.