missing translation for 'onlineSavingsMsg'
Learn More

Mycophenolate Mofetil N-Oxide (EP Impurity G), TRC

Product Code. 30133157
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133157 100 mg 100mg
30012806 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133157 Supplier Toronto Research Chemicals Supplier No. TRCM831470100MG
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 224052-51-1
Molecular Formula C23 H31 N O8
Molecular Weight (g/mol) 449.49
Synonym 4-Hexenoic acid, 6-(1,3-dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-, 2-(4-oxido-4-morpholinyl)ethyl ester, (4E)- (9CI), 2-(Morpholin-4-yl)ethyl (4E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoate N-Oxide, Mycophenolate Mofetil N-Oxide
IUPAC Name 2-(4-oxidomorpholin-4-ium-4-yl)ethyl (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoate
SMILES COc1c(C)c2COC(=O)c2c(O)c1C\C=C(/C)\CCC(=O)OCC[N+]3([O-])CCOCC3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C23H31NO8/c1-15(5-7-19(25)31-13-10-24(28)8-11-30-12-9-24)4-6-17-21(26)20-18(14-32-23(20)27)16(2)22(17)29-3/h4,26H,5-14H2,1-3H3/b15-4+
Formula Weight 449.205
Percent Purity >95%
Chemical Name or Material 2-(Morpholin-4-yl)ethyl (4E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoate N-Oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.