missing translation for 'onlineSavingsMsg'
Learn More

Mycobacterium Tuberculosis ESAT6 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18320842 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320842 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320842 Supplier Novus Biologicals Supplier No. NBP309114AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Mycobacterium Tuberculosis ESAT6 Polyclonal antibody specifically detects Mycobacterium Tuberculosis ESAT6 in Bacteria samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycobacterium Tuberculosis ESAT6
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 6 kDa early secretory antigenic target, 6 kDa early secretory antigenic target ESXA, ESAT 6, esaT6, ESXA
Host Species Rabbit
Immunogen EsaT-6 (Rv3875) of Mycobacterium tuberculosis
Purification Method Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.