missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium Tuberculosis CFP10 Antibody (KFB42), DyLight 405, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Mycobacterium Tuberculosis CFP10 Monoclonal antibody specifically detects Mycobacterium Tuberculosis CFP10 in Bacteria samples. It is validated for ELISA, Sandwich ELISA
Specifications
Specifications
| Antigen | Mycobacterium Tuberculosis CFP10 |
| Applications | ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | KFB42 |
| Conjugate | DyLight 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 10 kDa culture filtrate antigen cfp10, Culture filtrate protein 10, ESAT 6 like protein esxB, esxB, lhp, mtsA10, Secreted antigenic protein MTSA 10, tuberculosis CFP10 |
| Host Species | Mouse |
| Immunogen | Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with recombinant protein CFP10 M. tuberculosis H37RV. |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?