missing translation for 'onlineSavingsMsg'
Learn More

Mycobacterium Tuberculosis CFP10 Antibody (KFB16), DyLight 550, Novus Biologicals™

Product Code. 18403745 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403745 0.2 mL 0.20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403745 Supplier Novus Biologicals Supplier No. NBP105466R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mycobacterium Tuberculosis CFP10 Monoclonal antibody specifically detects Mycobacterium Tuberculosis CFP10 in Bacteria samples. It is validated for ELISA, Dot Blot, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycobacterium Tuberculosis CFP10
Applications ELISA, Dot Blot, Sandwich ELISA
Classification Monoclonal
Clone KFB16
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias 10 kDa culture filtrate antigen cfp10, Culture filtrate protein 10, ESAT 6 like protein esxB, esxB, lhp, mtsA10, Secreted antigenic protein MTSA 10, tuberculosis CFP10
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with recombinant protein CFP10 M. tuberculosis H37RV.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.